VWR® Sterile Opaque Sample Bags
Proveedor: Avantor
VWR®
EPN-4590-VW1 EPN-4590-VW1
93000-724CS 7246.42 MXN
CA93000-724 93000-724
VWR® Sterile Opaque Sample Bags
Bolsas Bolsas para muestras
Made of black-pigmented resin, sterile bags protect photosensitive samples such as vitamins, chemicals, pharmaceuticals, food supplements, plant extracts, and biological tissues.
  • Sterile
  • Opaque
  • Thickness accommodates abrasive chemicals
  • LDPE tubing
  • Write-on strip
  • Safety tab closures
Ideal for transport or long-term storage in food, pharmaceutical, and environmental industries; 3 mm thickness can accomodate abrasive chemicals.
Bags comply with EPA, FDA, USDA, and HACCP regulatory and industry requirements. RNase-, DNase-, and pyrogen-free.
Bags feature FDA-approved low-density polyethylene tubing, write-on strip, and safety tab closures.
Información para pedidos: Bags are packaged 500 per pack, 2 packs per case.