API 50 CHB/E Medium, bioMérieux
Proveedor: bioMérieux
50430
97060-444BX 2130.2 MXN
97060-444
API 50 CHB/E Medium, bioMérieux
Medios Medios preparados
API 50 CHB/E Medium is intended for the identification of Bacillus and related genera, as well as Gram-negative rods belonging to the Enterobacteriaceae and Vibrionaceae families. It is a ready-to-use medium which allows the fermentation of the 49 carbohydrates on the API 50 CH strip to be studied.