pH Test Strips, VWR Chemicals BDH®
Proveedor: Avantor
BDH®
92150.2047 92118.2047 92180.2047 92115.2047 92122.2047 92130.2047 92140.2047 92120.2047 92125.2047 92110.2047 92160.2047 92190.2047 92170.2047
BDH35311.604CS 16140.43 MXN
BDH35311.604 BDH83931.601 BDH35316.601 BDH83932.601 BDH35315.607 BDH35317.604 BDH35309.606 BDH83930.601 BDH83933.601 BDH35314.604 BDH35310.601 BDH35318.607 BDH35312.607
pH Test Strips, VWR Chemicals BDH®
Papeles de pruebas Tiras indicadoras de pH
Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use.
  • Designed for safe testing of dangerous, poisonous, or aggressive liquids
  • Long handle protects user from contact with sample
  • Fixed dye cannot bleed into the sample, protecting it against contamination and enabling measurements even in weakly buffered or strongly alkaline solutions
Dimensions: (LxW) 85x6 mm.
Información para pedidos: Strips are packaged 100 to a box. Included color chart has 4 different color blocks for each pH value