Titanium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Proveedor: Avantor
VWR Chemicals BDH® ARISTAR®
MSTI-10PPM-125ML CABDH89800-518 BDH82026-078 BDH89800-516 CABDH89800-188 BDH89800-518 BDH89800-188 MSTI-100PPM-125ML CABDH89800-516 BDH82026-252 BDH82026-250 CABDH82026-078
BDH82026-078EA 1410.26 MXN
CABDH89800-518 BDH89800-516 CABDH82026-250 CABDH89800-516 BDH82026-252 CABDH82026-078 CABDH82026-252 BDH82026-250 BDH82026-078 BDH89800-188 CABDH89800-188 BDH89800-518
Titanium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Normativas Estándares catiónicos Estándares de titanio
Used with ICP-MS, ICP-OES, DCP, GFAA, and AA
  • Long shelf life
  • NIST traceable
  • Precise and accurate
These ICP and ICP-MS CRMs are engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO Guide 34 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based on detailed error budgets, and trace metallic impurities (TMI) are determined by ICP and ICP-MS for single element CRMs
Each single element ICP standard ships with a Certificate of Analysis (C of A) and Safety Data Sheet (SDS). Specific lot C of A’s can also be found online.
Enhanced packaging extends the shelf life to 4 years by sealing the product in an external aluminized bag over the regular poly bottle.
This specially designed outer bag prevents transpiration (the loss of water, or solvent, through the container.) Therefore, most of these standards have a 4 year shelf life until the outer bag is opened. After opening the outer bag, the customer will mark the inner bottle with a one-year shelf life from that date.
Single element Certified Reference Materials (CRMs) are available in the BDH range for most of the periodic table, in concentrations ranging from 10, 100, 1,000, and 10,000 µg/mL
Please note: 1000 ppm = 1000 µg/mL