Hose Clamps, Adjustable, Stainless Steel
Proveedor: Avantor Fluid Handling
Masterflex®
WDC-36HS-10 WDC-16HS-10 WDC-72HS-10 WDC-64HS-10 WDC-24HS-10 WDC-28HS-10 WDC-88HS-10 WDC-48HS-10 WDC-104HS-10 WDC-20HS-10 WDC-3504S-10
MFLX06403-14EA 879 MXN
MFLX06403-14 MFLX06403-25 MFLX06403-15 MFLX06403-16 MFLX06403-21 MFLX06403-22 MFLX06403-11 MFLX06403-12 MFLX06403-23 MFLX06403-24 MFLX06403-13
Hose Clamps, Adjustable, Stainless Steel
Abrazaderas Pinzas para tubos
Ideal for securely clamping tubes in place.
  • Resists corrosion
The band-type worm drive clamp features stainless steel strap and screw housing with a stainless steel screw.