Masterflex® Hose Clamps, Acetal Copolymer, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
SNP-08 SNP-01 SNP-06 SNP-01 OHS SNP-12 SNP-22 SNP-24 SNP-34 SNP-19 SNP-10 SNP410C3 MAKE-UP SNP-18 SNP-14 SNP-1.5 SNP-1.25 WHITE SNP-03 WHITE SNP-28 SNP-02 SNP-32
MFLX06832-08CS 45.75 MXN
MFLX06832-08 MFLX06832-18 MFLX06832-22 MFLX06832-99 MFLX06832-10 MFLX06832-32 MFLX06832-24 MFLX06832-02 MFLX06832-01 MFLX06832-12 MFLX06832-34 MFLX06832-04 MFLX06832-14 MFLX06832-03 MFLX06832-06 MFLX06832-28 MFLX06832-20 MFLX95614-00 MFLX95613-00 MFLX95614-01
Masterflex® Hose Clamps, Acetal Copolymer, Avantor®
Abrazaderas Pinzas para tubos
Reusable white acetal copolymer clamps will not rust or corrode.
  • Resists corrosion
Recommended for continuous use from ambient temperature to 200ºF (93ºC). Clamps are nonconducting and stable in a wide variety of chemicals.
Once set, clamps stay put. To use, just position clamp on hose or tubing bundle and press the jaws together with a pair of pliers.