Masterflex® Sanitary Clamps, Nylon, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
31201-93 31201-95 31201-99 31201-97
MFLX31201-93EA 710 MXN
MFLX31201-93 MFLX31201-99 MFLX31201-97 MFLX31201-95
Masterflex® Sanitary Clamps, Nylon, Avantor®
Abrazaderas Pinzas para tubos
Compatible with metal and plastic Tri-Clamp® fittings.
  • Excellent chemical resistance
  • Rated to 150 psi
  • Sterilizable via Autoclave and Gamma Irradiation
  • Features anti-tamper zip-tie lock out
  • Manufactured and packaged in cleanroom
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.