Masterflex® I/P® Stainless-Steel IP66 Digital Dispensing Pump System, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
77715-26 77715-26 77715-26 77715-26 77715-26 77715-26 77715-26 77715-26 77715-26
MFLX77715-26-EUCS 0 MXN
MFLX77715-26-EU MFLX77715-26 MFLX77715-26-IL MFLX77715-26-AU MFLX77715-26-CH MFLX77715-26-IN MFLX77715-26-UK MFLX77715-26-DK MFLX77715-26-IT
Masterflex® I/P® Stainless-Steel IP66 Digital Dispensing Pump System, Avantor®
Bombas Bombas peristálticas
Dispense by Volume in mL (0.001 to 99999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle).
  • Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense
  • View pump performance continually
  • Antidrip function ensures dispensing accuracy
  • Open head sensor
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size.
Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction
Certificaciones: ETL, cETL, CE marked.
Información para pedidos: Includes NEMA 3-prong ground power cable. Accessories not included.