Chloride IC Standard, 1 mL = 1 mg Cl-, 1000 ppm Cl- (0.0282 Normal), Ricca Chemical Company
Proveedor: RICCA CHEMICAL CO
Chloride Standard, 500mL^ 1 mL = 1 mg Cl-, 1000 ppm Cl- (0.0282 Normal) (NaCl in water) (non-regulated). For ion chromatography. Clear, colorless, odorless. NIST traceable. Poly Bottle. 500mL