Cobaltous Chloride CS, USP / EP Red Primary Solution, 1 mL = 59.5 mg CoCl2.6H2O, Ricca Chemical Company
Proveedor: RICCA CHEMICAL CO
Cobaltous Chloride CS, 120mL^ USP Colorimetric Solution / EP Red Primary Solution, 1 mL = 59.5 mg CoCl2. 6H2O. Amber glass bottle. 120mL