Color Standard, APHA / Hazen (Platinum-Cobalt), Ricca Chemical Company
Proveedor: RICCA CHEMICAL CO
Color Standard, 500mL^ APHA / Hazen (Platinum-Cobalt), Color = 15 units. APHA, ASTM, ACS, EPA. Stock platinum-cobalt formula. Amber glass bottle. 500mL.