Nitrate Standard, 1 mL = 1 mg NO3-, 1000 ppm NO3- (226 ppm N), Ricca Chemical Company
Proveedor: RICCA CHEMICAL CO
Nitrate IC Standard^ 1000 ppm NO3 (KNO3 in water) (non-regulated). For ion chromatography. Clear, colorless. NIST traceable. 120mL.