Iron AA Standards, 1 mL = 1 mg Fe (1,000 ppm Fe)Fe in 3% HCl, RICCA Chemical Company
Proveedor: RICCA CHEMICAL CO
Iron Ion AA Standard^ 1000 ppm Fe ion in dilute HCl. For use in standardizing atomic absorption spectrophotometers. 100mL.