Just PROVE it
Simplifique el análisis de agua con un nuevo tipo de espectrofotómetros.
New range of instruments designed to meet all your requirements for water analysis - waste water, process water or drinking water and beverages.
Request Demo/Quote- Live ID, a new, data-rich 2D barcode on all cell and reagent tests for recognizing methods and transfering “live” data like expiry, lot number together with automatic, complementary calibration updates
- Cell Test Port - Direct insertion of round cell tests without opening any lid
- AQA excelente: verificación y documentación completas de todas las fases del control de calidad
- Easy cleaning - Cell holder removable
- Matrix Check easily detects interferences
Spectroquant® PROVE 100 | Spectroquant® Prove 300 | Spectroquant® Prove 600 |
Routine applications | Sensitive measurements | Complex analysis |
The best choice for use with cell tests and Vis measurements | Long lasting Xenon lamp makes it suitable for intensive use in UV-Vis | High-end UV-Vis optics and cuvettes up to 100 mm. Excellent resolution and sensitivity |
1.73016.0001 | 1.73017.0001 | 1.73018.0001 |