Stenner Low-Volume Compact Peristaltic Pump, 80 psi, 70 ml/hr; 120 V/60 Hz
MFLX74209-00
: Avantor
CE Compliant
- Maximum Flow Rate:0 L/h (0.036 gal./h)
- Power:120 VAC; 60 Hz
- Accuracy:±2%
- Max. pressure:5.5 bar (80 psi)
- Min. flow rate:0.07 L/h (0 gal./h)
- Description:Low-Volume Compact Peristaltic Pump, 80 psi, 70 ml/hr; 120 V/60 Hz
- Environmentally Preferable:
- Wetted materials:Santoprene, PE, PP, PVC, and ceramic
- Duty cycle continuous:Yes
- Run dry:Yes
- Self-priming:Yes
- Max. temperature:40 °C (104 °F)
- Warranty:1 year
- Cat. no.:MFLX74209-00