American Machine & Tool Self-Priming Centrifugal Pump, 316 SS, 100 GPM, 2 hp; 230/460 VAC
MFLX70620-22
: Avantor
Chamber in front of pump housing enables self-priming operation.
- Self-cleaning, semi-open impellers
These American Machine & Tool 316 stainless steel self-priming centrifugal pumps feature a continuous duty TEFC motor. Pumps are constructed of a 316 stainless steel impeller and carbon graphite and ceramic bearing. Models 70620-00, -02, -10. -12, -22, and -32 feature a Viton® o-ring and seal. Models 70620-72 and -80 feature a PTFE o-ring and seal.
- Maximum Flow Rate:378.5 L/min (100 gal./min)
- Power:230/460 VAC; 6/3 A; 3 phase
- Max. pressure:5.2 bar (75 psi)
- Description:Self-Priming 316SS Centrifugal Pump; 100 gal./min, 2 hp, 230/460 VAC
- Height:254 mm
- Length:45.7 cm
- Environmentally Preferable:
- Wetted materials:316 SS impeller; carbon graphite/ceramic bearing; Viton® O-ring & seal
- Duty cycle continuous:Yes
- Inlet connection:1.5 NPT(F)
- Motor hp:2
- Outlet connection:1.5 NPT(F)
- Motor type:TEFC
- Max. temperature:93 °C (200 °F)
- Warranty:1 year
- Cat. no.:MFLX70620-22
- Max. Head:29 m (95 ft.)
- Width:10.2 cm