VWR® Certified Precision Verification Thermometers
Proveedor: VWR
60207-0111 60207-0011 602070011 60207-0211
89095-698CS 6561.28 MXN
CA89095-696 89095-698 89095-696 89095-700
VWR® Certified Precision Verification Thermometers
Termómetros Analogue Thermometers
Used to verify sample temperatures in freezers, refrigerators, or incubators with traceability to NIST.
  • Organic liquid
  • Each item features individual serial numbers and a Statement of Accuracy indicating accuracy traceable to NIST
Tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST and DKD/PTB.
Each item features individual serial numbers and a Calibration Report stating corrections to one-tenth of the smallest division. Organic liquid.