VWR® Specific Gravity/Relative Density Plain Form Hydrometers, Traceable to NIST
Proveedor: VWR
61801-4611 61801-3511 61801-5611 61801-5911 61801-6011 61801-5111 61801-5811 61801-4211 61801-4911 61801-3711 61801-5011 61801-7211 61801-4511 61801-7811 61801-7011 61801-7711 61801-7411 61801-7311 61801-4311 61801-4411 61801-5311 61801-4811 61801-5211 61801-3611 61801-7611 61801-5711 61801-4111 61801-5511 61801-7511 61801-4711 61801-5411 61801-6111 61801-7111
34620-158CS 6922.08 MXN
34620-158 34620-178 34625-633 34620-156 34629-011 34623-106 34623-208 34622-205 34620-172 34620-170 34622-103 30617-754 34620-176 34622-001 34620-174 34615-006 34623-150 34629-601 34623-252 34620-168 34622-250 34625-600 34625-666 34620-006 34620-162 34620-160 34622-158 34620-100 34620-166 34620-164 34620-180 34623-300 34610-002
VWR® Specific Gravity/Relative Density Plain Form Hydrometers, Traceable to NIST
Hidrómetros
These plain form hydrometers are available in a variety of models measuring specific gravity/relative density (g/cm³) of liquids lighter and heavier than water in various ranges from 0.690 to 3.000.
  • Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60 °F (15 °C)
Plain form hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
Certificaciones: Hand inspected by our U.S. quality team, individually serialized.
Envase: Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.