VWR® Oven Thermometers
Proveedor: VWR
608030511 608031111 60803-0911 60803-0511 60803-0711 60803-1111 608030711 608030911
89095-818CS 2408.28 MXN
CA89095-814 89095-816 CA89095-812 89095-814 CA89095-818 CA89095-816 89095-818 89095-812
VWR® Oven Thermometers
Termómetros Analogue Thermometers
Oven thermometers are individually serialized, and partial immersion versions feature a round glass swell to hold the thermometer in place.
  • Individually serialized
  • Lead-free white glass
  • Red organic liquid
Individually packaged in recyclable packaging with non-roll fitting.