Open Ring Support, Ace Glass
Proveedor: Ace Glass
11171-17 11171-22 11171-12
89075-682EA 1960.66 MXN
89075-682 89075-684 89075-680
Open Ring Support, Ace Glass
Soportes Soportes de matraces
This Flexaframe open ring support can be held by conventional laboratory clamp holders
Use it to support equipment such as leveling bulbs and separatory funnels from Flexaframe. Three different sizes are available, all manufactured from aluminum.