Rotating Bottle Holder, PVC, TrippNT
Proveedor: TrippNT
50087
10012-842EA 4550.74 MXN
10012-842
Rotating Bottle Holder, PVC, TrippNT
Gradillas Gradillas de botellas
White PVC rotating bottle holder stores 8 bottles of 500mL and 1 bottle of 1000mL adds safety and convenience by storing top heavy bottles in this stabilizing and rotating nine-bottle holder
Made of heavy-duty PVC, it holds eight 500mL and one 1L wash or squeeze bottles. Middle holder can accommodate ninth wash bottle or hold a beaker to collect drips from other bottles. Secure storage for a large numbers of bottles with easy access by rotation. Makes storage easier in cabinets or bench top for sharing resources.