SMITH & WESSON® ELITE™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Proveedor: Kimberly-Clark
Smith & Wesson®
21303 21302 21306
89215-600CS 3828.83 MXN
89215-602 89215-598 89215-600
SMITH & WESSON® ELITE™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Gafas
Lightweight 'retro-style' design provides the latest fashion statement.
  • Wide temple design ensures structural integrity and stability
  • Anti-scratch dual lenses with an 8-base curve provide wrap-around protection
  • High performance polycarbonate lens provides 99.9% UV protection
Weight: 59 g (0.13 lbs.).
Certificaciones: Meets ANSI Z87.1+ standards.
Información de suministro: Supplied with micro-fiber carry pouch that doubles as a cleaning cloth.