Combination Heating Mantle, Ace Glass Incorporated
Proveedor: Ace Glass
12061-128 12061-123 12061-05 12061-71 12061-102 12061-65 12061-09 12061-31 12061-11 12061-61 12061-63 12061-03 12061-113 12061-07 12061-73 12061-117 12061-21 12061-137 12061-13 12061-134 12061-19 12061-115 12061-108 12061-140 12061-67 12061-75 12061-29 12061-77
89076-118EA 107870.13 MXN
89076-130 89076-152 89076-150 89076-118 89076-138 89076-116 89076-114 89076-136 89076-158 89076-112 89076-156 89076-134 89076-154 89076-110 89076-132 89076-140 89076-160 89076-108 89076-128 89076-106 89076-126 89076-148 89076-146 89076-124 89076-144 89076-122 89076-120 89076-142
Combination Heating Mantle, Ace Glass Incorporated
Calentadores Capas calefactoras
The lower profile, safer, multi-place Combo Mantle allows the user to easily replace an element in a matter of minutes, should that element fail
Mantles are available as a six-place unit for 100–300mL flasks, or a three-place unit for 500–1000mL flasks. The clear, anodized aluminum cabinet has a black PTFE-coated stainless steel top and comes complete with a glassware superstructure consisting of two upright rods, two horizontal rods, and four double-sided, open faced clamps. The six-place mantle features six spring-type glassware clamps, and the three-place model features three.
A choice of two 3-place controllers are available; a percentage-timer version that pulses full-line voltage to each position according to dial setting, or a proportional voltage version that supplies a constant, steady-state voltage to each position. Controllers are supplied with 1.8m (6') multi-conductor interconnect cord to mantle and one 1.2m (4') long, three-wire power cord with grounding plug.
Note: The six-place mantle uses two 3-place controllers.
By unscrewing two captive screws and lowering the front panel of the cabinet, the disposable element containers can be replaced by detaching the two lead wires and sliding the containers out. A new unit can be installed by reversing the procedure. These elements act as spill containment chambers so a spill does not damage other parts of mantle.
Información para pedidos: Select appropriate mantle based upon desired flask size/type and power needed, then select type and number of required controller(s)