RF20 Portable Brix Refractometer with Automatic Temperature Compensation, Extech®
Proveedor: Extech
RF20
89370-226EA 3817.8 MXN
89370-226
RF20 Portable Brix Refractometer with Automatic Temperature Compensation, Extech®
Refractómetros Refractómetros de mano
Portable Salinity Refractometer (0–100ppt) with ATC that measures the concentration of dissolved salt in fluids
The RF20 provides accurate and repeatable measurements on an easy-to-read scale. Requires only 2 or 3 drops of solution. It features Automatic Temperature Compensation (ATC) from 10 to 30°C. Compact size, easy to operate.
Certificaciones: CE marked.
Información para pedidos: Supplied complete with case, calibration screwdriver, and calibration solution. Refractometer also includes manufacturer's one-year warranty.