Auto-Desiccator Cabinets, Ace Glass
Proveedor: Ace Glass
6257-50 6257-64
89054-070EA 62342.94 MXN
89054-070 89054-072
Auto-Desiccator Cabinets, Ace Glass
Desecadores Armarios desecadores
This stackable, plastic Secador™ desiccator cabinet is perfect for cramped locations
Can be safely stacked up to two high. Cabinet offered clear or blue with clear door. With patented dehumidifying module—automatic desiccant regeneration and built-in hygrometer. Unit has three shelves, four shelf positions. Patented sealed construction—for air-tight, dust- and moisture-free storage. Two latches; provision for lock or tamper evident seal.