DEHA Reagent 1 Powder Pillows, Hach
Proveedor: Hach
2167969 2167969
97026-654EA 47.45 MXN
97026-654 CA21679-69
DEHA Reagent 1 Powder Pillows, Hach
Kits para test medioambiente
For determination of oxygen scavengers by the iron reduction method. 
Hach method 8140 determines carbohydrazide, diethylhydroxylamine (DEHA), hydroquinone, iso-ascorbic acid (erythorbic acid) and methylethylketoxime.
Información para pedidos: Pack of 100 Powder Pillows.