Lattice Rod, Aluminum
Proveedor: EISCO SCIENTIFIC LLC
CH0658PLM3 CH0658PLM6
470140-276EA 351.92 MXN
470140-276 470140-274
Lattice Rod, Aluminum
Varillas de retorta
Resists corrosion
  • Made of T6 Temper 6063 aluminum alloy
These anodized, smooth, unthreaded aluminum rods are used for constructing lattice rod assemblies.