Ozone AccuVac® Ampules, MR, Hach
Proveedor: Hach
2517025 2517025
97026-172EA 37.11 MXN
97026-172 CA25170-25
Ozone AccuVac® Ampules, MR, Hach
Kits para test medioambiente
For determination of Ozone by the Indigo method, using AccuVac® Ampules.
  • Fast
  • Easy
  • Accurate
  • Use them anywhere you need results.
Información para pedidos: Pack of 25 ampules.