SpecCheck Secondary Gel Standards Set, DPD Chlorine, LR, Hach
Proveedor: Hach
SpecCheck
2635300 2635300
97026-500EA 11500 MXN
CA26353-00 97026-500
SpecCheck Secondary Gel Standards Set, DPD Chlorine, LR, Hach
Normativas Estándar par análisis medioambiental
DPD Chlorine, Low Range, 0-2.00 mg/L as Cl2.
  • Fast and Convenient
  • Stable
Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.
Información para pedidos: Set of 4 jars.