sensION Gel Filled Combination pH Electrode, 5-pin connector, Hach
Proveedor: Hach
5193500 5193500
97021-582EA 12154.15 MXN
97021-582 CA51935-00
sensION Gel Filled Combination pH Electrode, 5-pin connector, Hach
Electrodos Electrodos pH
Standard combination pH probe with a non-refillable gel reference and built-in temperature sensor.
  • Accurate
  • Easy to Use
  • Rugged and Durable
  • Flexible
Contains fixed cable with 5-pin circular connector compatible with Hach sensION meters.