Sample Cells, Round Glass, Hach
Proveedor: Hach
2084900 2084900 2427606 2401906 2427606 2401906
97026-770EA 48.7 MXN
97026-770 97026-754 CA24019-06 CA24276-06 CA20849-00 97026-768
Sample Cells, Round Glass, Hach
Cubetas Cubetas para espectrofotómetros
Round, glass sample cells for use with Hach Spectrophotometers
Cells come in pack of six with caps.