High Capacity Force Gauge with PC Interface, Extech
Proveedor: Extech
475055-NIST 475055
97049-994EA 26505.86 MXN
97049-994 10171-884
High Capacity Force Gauge with PC Interface, Extech
Dinamómetros
High capacity force gauge with PC Interface and optional software
Features a large LCD with back light feature and Positive/Reverse display for easy readability.
Certificaciones: CE marked.
Información para pedidos: 1-Year Warranty. Complete with tension and compression adaptors, six AA 1.5V batteries, and case.