pH Buffer Solution Kits, Color-Coded, pH 4.01, 7.00 and 10.01, Hach
Proveedor: Hach
2507300 2507200 2507200 2947600
97021-450EA 6800 MXN
97021-450 CA97021-450 97021-452 97021-448
pH Buffer Solution Kits, Color-Coded, pH 4.01, 7.00 and 10.01, Hach
Tampones Tampones pH
Convenient 3-packs of color-coded buffer solutions offer a 33% cost reduction over single bottle purchase
Each pack includes one container each of pH standardization buffer solutions at pH 4.01 (red), 7.00 (yellow), and 10.01 (blue).
They are recommended for automatic calibration and assure accuracy (±0.02 pH units).
Certificaciones: Traceable to NIST.