VWR® V5 Microscope Camera
Proveedor: VWR, part of Avantor
1100600400011
89404-504EA 27678.43 MXN
89404-504
VWR® V5 Microscope Camera
This easy-to-use still and video digital 5 megapixel camera is adaptable to a variety of applications.
  • Easy to use
  • Adaptable
  • Quality high-resolution live camera feed
The VWR® attachable microscope camera offers 5.0MP live resolution and is equipped with a USB cable. It includes Images Plus 2.0 Software for PC and MAC.
Certificaciones: C-UL, UL Listed. CSA certified.
Información para pedidos: Microscope camera is supplied with the accessories listed above and one-year warranty.