VWR® Stereo Zoom Trinocular Microscope
Proveedor: VWR
VWR®
1101001400723V 1101001700272V 1101001400932V 1100200500501 1101001206431 1101000600531V 1101002400372V 1101001400962V 1101001700332V 1101001700302V 1101001000232V 1101001000241V 1101001400852V 1101001400641V 1101000900121V 1101001400702V 1101000600431V 1101001700312V 1101006400111V 1101002300281V 1101001700282V 1101001400733V 1101001400952V 1101001400882V 1101001700292V 1101002300322V
89404-476EA 57391.36 MXN
89403-416 89403-438 89403-436 89403-458 89403-414 89403-418 89403-420 89403-464 89403-442 89403-440 89403-468 89403-424 89403-446 89403-444 89403-466 89403-422 89403-460 89404-476 89403-426 89403-430 89403-452 89403-450 89403-456 89403-432 89403-454 89403-470
VWR® Stereo Zoom Trinocular Microscope
Microscopios
High performance Stereo Microscope is ideally suited for life science, industrial and pharmaceutical inspection, electronic assembly, and bonding operations.
  • Trinocular stereo microscope head
  • Widefield eyepieces
  • Transmitted illumination
The VWR® Stereo Zoom Microscope features the Greenough Zoom Optical System, a trinocular head with 360° rotation, photo/video: 100% [right eyetube], and widefield eyepieces 10X/F.N. 23. The unit offers 6.7:1 zoom ratio, WD = 113mm, and a magnification range of 0.75X–5X. Additional standard features include incident/transmitted large working area stand, a 12V/10W intensity-controlled halogen incident, and transmitted illumination. Main power supply is 100–240V.
Información para pedidos: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.