Air Sampling System, C.A.R.B. Octopus Glass, Ace Glass
Proveedor: Ace Glass
7489-22 7489-26 7489-20 7489-24 7489-08 7489-04
89084-284EA 13823.67 MXN
89084-290 89084-288 89084-286 89084-284 89084-294 89084-292
Air Sampling System, C.A.R.B. Octopus Glass, Ace Glass
Muestreadores Muestreadores de aire
These vertical systems fit more easily into air sampling stations, trailers and cabinets, and provide a more efficient methodology of sample collection
Both the 4-port and 8-port manifolds are available with either a glass sampling cane with an inverted glass cone, or with a straight glass cane with a polyethylene inverse funnel on top. Approximate I.D. of the sampling tube is 25 mm (1 inch). Approximate overall height of four-port system is 150 cm (60 inches). Approximate overall height of eight-port system is 155 cm (62 inches). Fabricated from borosilicate glass. (C.A.R.B. = California Air Resources Board.)
All the inner surfaces that are exposed to the air are either inert borosilicate glass or inert PTFE. Thanks to the convenient Ace-Threds®, the systems can be safely and easily disassembled for cleaning or replacing parts. Both 4-port and 8-port canes offer a PTFE sampling port outside for spot sampling or EPA auditing. The heavy duty #7 Ace-Thred ports on the manifolds can be fitted to any type of tubing, and can be sealed with ferrules and bushings to facilitate connection to air sampling instrumentation and monitors.
Información accesorios: Economical air sampling cane eliminates the bend and the funnel. The tube length for all canes is 120 cm (471/4")."
Información para pedidos: All complete systems include sampling cane/hood (89083-166 curved glass with glass funnel top; of 89083-170 straight glass with polyethylene funnel) and 4- or 8-port glass manifold, plus 89059-202 collection bottle (250 mL); 89083-162 nylon #7 bushing with CAPFE o-ring; 89084-370 PTFE plug stopper (#7) with CAPFE o-ring; 89051-124 "ferrule design" nylon bushing (#7) with 7.5 mm center hole; and 89075-734 pre-drilled ferrules (#7 size, pack of 12).