NitriVer® 3 Nitrite Reagent Powder Pillows, 25 ml, Hach
Proveedor: Hach
1406528 1406599 1406599
97026-586PK 3100 MXN
97026-586 10765-226 CA14065-99
NitriVer® 3 Nitrite Reagent Powder Pillows, 25 ml, Hach
Kits para test medioambiente
For low range nitrite determination by the NitriVer® 3 Diazotization Method: EPA Compliant - Hach Method 8507
Range: up to 0.300 mg/L NO2-N Sample Size: 25 mL.
Información para pedidos: Pack of 100 powder pillows.