sensION Platinum Series Combination pH Electrode, Flat End, BNC Connector, Hach
Proveedor: Hach
5191500 5191500
97021-590EA 21321.55 MXN
CA11014-346 97021-590
sensION Platinum Series Combination pH Electrode, Flat End, BNC Connector, Hach
Electrodos Electrodos pH
Hach Platinum Series pH probe is a standard combination pH probe with a replaceable gel reference
  • Accurate
  • Easy-to-use
  • Rugged and durable
  • Flexible
Probe comes with two electrolyte cartridges and has a fixed cable with a BNC connector. It has a flat end that requires minimal immersion and smaller sample volumes, and can be used for surface measurements in water quality applications.