Stablcal® Turbidity Standards, 1.0 to 40 NTU, Hach
Proveedor: Hach
Stablcal®
2659853 2659942 2659842 2659849 2659842 2746353 2660149 2660153 2746356 2660153 2660142 2659942 2746353 2660149 2746356 2659949 2659849 2659949 2660142
97026-436EA 5200 MXN
97026-436 97026-458 97026-502 CA11016-416 97026-416 CA27463-53 CA71002-460 CA26599-49 89231-162 CA11016-428 CA2659849 97026-472 89231-164 CA11016-440 97026-452 97026-166 97026-302 CA11016-432 CA26601-49
Stablcal® Turbidity Standards, 1.0 to 40 NTU, Hach
Normativas Estándar para turbidedad
Stablcal® Turbidity Standards for all turbidimeters.
  • Precise, premixed formazin solutions
  • Based on formazin primary standards
  • Enhanced stability saves time and labor
  • Confidence in results
USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter