FerroMo™ Iron Reagent Set, Hach
Proveedor: Hach
2544800 2544800
97026-982EA 8400 MXN
97026-982 CA25448-00
FerroMo™ Iron Reagent Set, Hach
Kits para test medioambiente
For determination of iron in cooling waters containing molybdate-based treatments
Hach Method 8365. Range: 0.01 - 1.80 mg/L Fe. Reagent Set Includes: FerroMo™ Reagent 1 Powder Pillows and FerroMo™ Reagent 2 Powder Pillows for 100 tests.