Ultrasonic Baths, Mechanical, Branson Ultrasonics
Proveedor: Branson
CPX-952-836R CPX-952-236R CPX-952-136R CPX-952-816R CPX-952-116R CPX-952-516R CPX-952-336R CPX-952-816R CPX-952-336R CPX-952-516R CPX-952-836R CPX-952-316R CPX-952-536R CPX-952-536R CPX-952-136R CPX-952-316R CPX-952-216R CPX-952-236R CPX-952-216R CPX-952-116R
89375-454EA 25957.8 MXN
89375-462 CA89375-450 89375-450 CA89375-462 89375-460 CA89375-452 CA89375-464 89375-466 89375-454 89375-464 CA89375-460 89375-452 CA89375-458 89375-458 89375-468 89375-456 CA89375-454 CA89375-466 CA89375-456 CA89375-468
Ultrasonic Baths, Mechanical, Branson Ultrasonics
Baños ultrasónicos
M Series baths feature 60 minute or continuous operation, sweep frequency which eliminates standing waves and creates consistent cavitation through the tank
Tranducers are 40KHz rugged industrial transducers. Controls are located above and behind the cleaning tank in an easily accessible control tower panel, and raised above the tank to avoid damage from cleaning solutions. This unit has convenient built-in pour drains on the .5 and .75-gallon models. Tank drains with valves are incorporated on models 1.5-gallon and larger.
Certificaciones: UL, CSA, CE, FCC, ISO9001, ISO14001.
Información para pedidos: Accessories are available; see accessory datasheet for more information.