Ultrasonic Baths, Digital, Branson Ultrasonics
Proveedor: Branson
CPX-952-119R CPX-952-219R CPX-952-539R CPX-952-819R CPX-952-839R CPX-952-139R CPX-952-839R CPX-952-239R CPX-952-519R CPX-952-119R CPX-952-339R CPX-952-539R CPX-952-819R CPX-952-519R CPX-952-239R CPX-952-319R CPX-952-319R CPX-952-219R CPX-952-339R
89375-432EA 19455.24 MXN
89375-440 CA89375-430 CA89375-442 89375-444 89375-432 89375-442 89375-430 89375-448 CA89375-436 CA89375-448 89375-436 89375-446 CA89375-438 89375-434 CA89375-432 CA89375-444 CA89375-434 CA89375-446 89375-438
Ultrasonic Baths, Digital, Branson Ultrasonics
Baños ultrasónicos
CPX Series features 99 minute timer or continuous operation, with degassing capability of up to 99 minutes ideal for sample preparation applications
Unit features sweep frequency which eliminates standing waves and creates consistent cavitation through the tank. Transducers are 40KHz rugged industrial transducers. Controls are located above and behind the cleaning tank in an easily accessible control tower panel, and raised above the tank to avoid damage from cleaning solutions. This unit has convenient built-in pour drains on the .5 and .75-gallon models. Tank drains with valves are incorporated on models 1.5-gallon and larger.
Ultrasonic power tracking allows the same ultrasonic power to be delivered to the tank, delivering unwavering cavitation, steady activity in the bath even when load conditions change. Operator can adjust the tank to use 100% of power or change to a lower amplitude of acoustic energy delivering 70% power for delicate applications if needed. Sleep mode goes into effect when cycle is complete and no control keys are touched for 15 minutes. Touching any key causes the unit to come back on when cycle is complete.
Certificaciones: UL, CSA, CE, FCC, ISO9001, ISO14001.
Información para pedidos: Accessories are available; see accessory datasheet for more information.