ECR Masking Reagent Solution, 25 ml, Hach
Proveedor: Hach
2380123 2380123
89231-062EA 1000 MXN
CA2380123 89231-062
ECR Masking Reagent Solution, 25 ml, Hach
Kits para test medioambiente
For Aluminum determination by the Eriochrome Cyanine R Method
Hach method 8326.