Cyanuric Acid 2 Reagent Powder Pillows, Hach
Proveedor: Hach
246066 246066
97026-542EA 27.85 MXN
97026-542 CA2460-66
Cyanuric Acid 2 Reagent Powder Pillows, Hach
Kits para test medioambiente
For determination of cyanuric acid by the melamine turbidimetric method
Hach method 8139. Sample size: 25 mL. Also used in Model CY-3 test kits.
Información para pedidos: Pack of 50 powder pillows.