Stablcal® Turbidity Standards, 100 to 7500 NTU, Hach
Proveedor: Hach
Stablcal® StablCal®
2584202 2660649 2660249 2660542 2660449 2660249 2660649 2660549 2660642 2660449 2660642 2660442 2660242 2660542 2660442 2660549 2584202 2660242
97026-474EA 5800 MXN
CA11016-424 CA11016-418 97026-396 CA11016-430 97026-474 97026-398 97026-454 97026-476 97026-498 CA11016-412 97026-456 CA11016-420 CA26602-49 CA25842-02 97026-390 CA26604-49 97026-392 CA26606-49
Stablcal® Turbidity Standards, 100 to 7500 NTU, Hach
Normativas Estándar para turbidedad
Stablcal® Turbidity Standards for all turbidimeters.
  • Precise, premixed formazin solutions
  • Based on formazin primary standards
  • Enhanced stability saves time and labor
  • Confidence in results
USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter