Ultimate Thermometer, Sper Scientific
Proveedor: Sper Scientific
800043C
97035-806EA 15879.53 MXN
97035-806
Ultimate Thermometer, Sper Scientific
Termómetros Termómetros digitales
Infrared, RTD and thermocouple capabilities in a single unit.
This unique thermometer combines Infrared, RTD and thermocouple capabilities in a single unit. With Ultimate Thermometer you’re certain to have a solutions for every temperature measurement application you encounter. For general purpose applications use any standard type K, J, R, E, or T thermocouple probe. For a very precise temperature reading, use the PT100 RTD probe included. To measure the temperature inside a furnace, just point the built-in IR thermometer, which features a laser guide and adjustable emissivity. Other sophisticated features include RS232 computer interface, C and F selection, min/max memory, relative temperature, auto power off, and hold. Has a tripod back and an easy to read back lit display with large 0.5" high digits.
Certificaciones: N.I.S.T. traceable certificate of compliance (for certified model only).
Información para pedidos: 5-year warranty. Supplied with a hard carrying case and a 9V battery.