Mantle, Spherical, Ace Glass Incorporated
Proveedor: Ace Glass
12031-25 12031-19 12031-23 12031-13 12031-17 12031-11 12031-07 12031-05 12031-21
89075-848EA 46655.87 MXN
89075-840 89075-832 89075-842 89075-834 89075-844 89075-836 89075-846 89075-838 89075-848
Mantle, Spherical, Ace Glass Incorporated
Calentadores Capas calefactoras
Fabric mantle covers both top and bottom of 1-, 2-, or 3-neck flasks
Temperature range is Ambient +10° to 450°C. The circumferential zipper holds the halves snugly together to prevent heat loss, and the side split in the top section allows for easy flask removal. Each half has a detachable 122cm (4') cord and interlocking connector. 230V, CE-approved versions are available.
Certificaciones: CSA certified.