REDISHIP Purifier® Logic®+ Class II A2 Biosafety Cabinets, Labconco®
Proveedor: Labconco
302589101 302589001 302519001 302689001 302319101 302619001 302389101 302489001 302689101 302389001 302489101 302319001 302419001 302519101 302419101 302619101
76317-626EA 339037.2 MXN
76317-634 76317-632 76317-628 76317-640 76318-336 76317-636 76317-614 76317-624 76317-642 76317-612 76317-626 76318-334 76317-616 76317-630 76317-622 76317-638
REDISHIP Purifier® Logic®+ Class II A2 Biosafety Cabinets, Labconco®
Vitrinas de gases y recintos Armarios de seguridad biológica y microbiológica
REDISHIP Purifier Logic+ Class II A2 Biosafety Cabinets use the same ECM blower technology as the Purifier Logic®, the Logic+ and use the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load.
In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
The cabinet provides personnel, product and environmental protection and is suitable for work in biosafety levels 1, 2 and 3. These biohazard cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection.
Logic+ Biosafety Cabinets excel in user comfort and functionality. Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, patented curved air grille with built in armrest, fully accessible controls and eye-level mounted color display with MyLogic™ operating system. MyLogic provides intuitive programming, color coded menus, status messages and alarms with diagnostic troubleshooting—all using full sentences.
Información para pedidos: All models are 115V and include UV light, service fixture, pass through, and sealed duplex receptacle (115V). Choose from an 8" or a 10" sash height. Some models include a telescoping base stand (shown in image). In other models, the stand is sold separately. Base stand requires assembly. Dimensions shown below are W×D×H. Please refer to product specifications for all details.