ABRATEC™ Abrasive Cloths, Contec®
Proveedor: Contec
ABRATEC™
ATQT1350 ATQT0280 ATQT0360
52428-876CS 8933.66 MXN
52428-876 52428-880 52428-878
ABRATEC™ Abrasive Cloths, Contec®
Toallitas Almohadillas de limpieza
Corrosion of stainless equipment in sterile process manufacturing is a significant problem for many life science manufacturers. Corroded surfaces are difficult to clean and disinfect because the rough surfaces do not allow good contact in wiping and provide places for bacteria to accumulate. This can result in sterility failures and lot rejections. Replacement equipment is a major capital expense and also represents significant loss of production due to facility downtime.
  • Various grit levels allow a wide range of remediation, including disinfectant residue removal
  • Quilted textile backing increases durability and abrasion resistance
  • Sealed edge reduces possible particulate contamination
  • No foam construction enables compatibility with a wide range of chemicals
  • Ideal for stainless steel remediation and minimizing rouging due to residues or soil
ABRATEC™ Abrasive Cloths are made with various levels of abrasive grits backed with the unique Quiltec® fabric. Quiltec® fabric is significantly more durable and abrasion resistant than similar foam-backed abrasives. This abrasion resistance and the sealed border technology allow the Quiltec® side to remove sanding debris without contributing additional particles to the remediation process.
ABRATEC™ Abrasive Cloths are designed to work with non-ketone based solvents, including the IPA/water blends typically found in sterile processing environments, as well as most commonly used cleanroom disinfectants. These cloths are also compatible with autoclave and gamma irradiation processing to help maintain the integrity of sterile environments. They are cleaner, have lower particulation, and are more chemically compatible than similar foam-backed abrasive products.