Order Entry
PIG® HazMat Spill Kit in 65-Gallon Overpack Salvage Drum, New Pig
PIG® HazMat Spill Kit in 65-Gallon Overpack Salvage Drum, New Pig
  56620-590
undefined
PIG® HazMat Spill Kit in 65-Gallon Overpack Salvage Drum, New Pig
  56620-590
 :  KIT343
$48,300.00
  Each (1UN)
  • Capacity:
    Up to 147.6 L (39 gal.)
  • Cat. no.:
    56620-590
  • Supplier No.:
    KIT343
Overpack drum container is UN Rated for shipping waste after spill cleanup
Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail. Lightweight, 100% polyethylene container resists chemicals and keeps contents clean and dry. Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole. Large, color-coded label makes identification fast and accurate. Ledges molded into container make kit easy to move by forklift. Tamperproof seals help prevent pilfering of spill response supplies.
No wheels. UN1H2/X200/S.
Lift-out, prepacked baskets speed access and guard contents from UV. PIG HazMat Socks stop spreading spills; PIG HazMat Pads and Pillows absorb quickly. PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide